missing translation for 'onlineSavingsMsg'
Learn More

Benzoyl-DL-valine 98.0+%, TCI America™

Catalog No. B040125G
Change view
Click to view available options
Quantity:
1 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B040125G 25 g
B04011G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B040125G Supplier TCI America Supplier No. B040125G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2901-80-6
Molecular Formula C12H15NO3
Molecular Weight (g/mol) 221.256
MDL Number MFCD00038283
InChI Key MIYQNOPLWKCHED-UHFFFAOYSA-N
PubChem CID 226682
IUPAC Name 2-benzamido-3-methylbutanoic acid
SMILES CC(C)C(C(=O)O)NC(=O)C1=CC=CC=C1

Specifications

Melting Point 132°C
Color White
Quantity 25 g
Formula Weight 221.26
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Benzoyl-DL-valine