missing translation for 'onlineSavingsMsg'
Learn More

Benzoyl chloride, 98+%, ACS reagent

CAS: 98-88-4 | C7H5ClO | 140.57 g/mol

Supplier:  Thermo Scientific Chemicals 402030025

 View more versions of this product

Catalog No. AC402030025

Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Benzoyl chloride
98.0
-1.0°C
198.0°C
0.005% max.
98+%
C7H5ClO
C6H5COCl
2.5 kg
01,50; 05,24; 13,219
15, 1115
Solubility in water: reacts, 2 mg/mL. Other solubilities: decomposes in alcohols and in water, soluble in ether, benzene; miscible with oils,, carbon disulfide, ccl4
C1=CC=C(C=C1)C(=O)Cl
140.57
0.0012 Pas (30°C)
140.57
ACS Reagent
98-88-4
100.0
1.2100g/mL
93°C
Authentic
Glass bottle
1.5520 to 1.5540
MFCD00000653
09, 182
1.21
benzenecarbonyl chloride, benzoic acid, chloride, benzoylchloride, alpha-chlorobenzaldehyde, benzoic acid chloride, benzoylchlorid, benzaldehyde, alpha-chloro, a-chlorobenzaldehyde, ccris 802, unii-vty8706w36
PASDCCFISLVPSO-UHFFFAOYSA-N
benzoyl chloride
7412
CHEBI:82275
98+%
Liquid

Chemical Identifiers

98-88-4
140.57
PASDCCFISLVPSO-UHFFFAOYSA-N
7412
benzoyl chloride
C7H5ClO
MFCD00000653
benzenecarbonyl chloride, benzoic acid, chloride, benzoylchloride, alpha-chlorobenzaldehyde, benzoic acid chloride, benzoylchlorid, benzaldehyde, alpha-chloro, a-chlorobenzaldehyde, ccris 802, unii-vty8706w36
CHEBI:82275
C1=CC=C(C=C1)C(=O)Cl
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
May cause an allergic skin reaction.
Harmful if swallowed.
Causes severe skin burns and eye damage.
Harmful in contact with skin.
Toxic if inhaled.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat

GHS Signal Word: Danger

EINECSNumber : 202-710-8

RTECSNumber : DM6600000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only