missing translation for 'onlineSavingsMsg'
Learn More

Benzotrifluoride 98.0+%, TCI America™

Catalog No. T043925G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T043925G 25 g
T0439500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T043925G Supplier TCI America Supplier No. T043925G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 98-08-8
Molecular Formula C7H5F3
Molecular Weight (g/mol) 146.112
MDL Number MFCD00000372
InChI Key GETTZEONDQJALK-UHFFFAOYSA-N
Synonym benzotrifluoride, trifluoromethyl benzene, alpha,alpha,alpha-trifluorotoluene, phenylfluoroform, benzene, trifluoromethyl, benzenyl fluoride, benzylidyne fluoride, usaf ma-16, a,a,a-trifluorotoluene, trifluoro toluene
PubChem CID 7368
ChEBI CHEBI:36810
IUPAC Name trifluoromethylbenzene
SMILES C1=CC=C(C=C1)C(F)(F)F

Specifications

Melting Point -29°C
Color Colorless
Boiling Point 102°C
Quantity 25 g
UN Number 2338
Formula Weight 146.11
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Benzotrifluoride
EINECSNumber (3)-0086
RTECSNumber XT9450000
TSCA Yes