missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzotrifluoride, 99%
Description
It is applied as an alternative solvent to dichloromethane, where it has a higher bp. It is used as an intermediate for dyes and pharmaceuticals, as a solvent and a dielectric fluid, a vulcanizing agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is applied as an alternative solvent to dichloromethane, where it has a higher bp. It is used as an intermediate for dyes and pharmaceuticals, as a solvent and a dielectric fluid, a vulcanizing agent.
Solubility
Soluble in water at 20°C0.45g/L.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
It is applied as an alternative solvent to dichloromethane, where it has a higher bp. It is used as an intermediate for dyes and pharmaceuticals, as a solvent and a dielectric fluid, a vulcanizing agent.
Solubility
Soluble in water at 20°C0.45g/L.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications
Specifications
| Melting Point | -29°C |
| Density | 1.188 |
| Boiling Point | 101°C to 103°C |
| Flash Point | 12°C (54°F) |
| Odor | Aromatic |
| Refractive Index | 1.4145 |
| Quantity | 1000 g |
| UN Number | UN2338 |
| Beilstein | 1906908 |
| Merck Index | 14,1110 |
| Show More |
Safety and Handling
| Hazard Category | H225 |
| Hazard Statement | GHS H Statement H225 Highly flammable liquid and vapour. |
| Precautionary Statement | P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: BENZOTRIFLUORIDE |
| EINECSNumber | 202-635-0 |
| RTECSNumber | XT9450000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only