missing translation for 'onlineSavingsMsg'
Learn More

Benzotrichloride 99.0+%, TCI America™

Catalog No. T039925G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T039925G 25 g
T0399500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T039925G Supplier TCI America Supplier No. T039925G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 98-07-7
Molecular Formula C7H5Cl3
Molecular Weight (g/mol) 195.47
MDL Number MFCD00000786
InChI Key XEMRAKSQROQPBR-UHFFFAOYSA-N
Synonym benzotrichloride, trichloromethyl benzene, alpha,alpha,alpha-trichlorotoluene, phenylchloroform, toluene trichloride, benzenyl trichloride, benzene, trichloromethyl, benzyl trichloride, benzylidyne chloride, phenyltrichloromethane
PubChem CID 7367
ChEBI CHEBI:82274
IUPAC Name (trichloromethyl)benzene
SMILES ClC(Cl)(Cl)C1=CC=CC=C1

Specifications

Melting Point -5°C
Color Yellow
Boiling Point 221°C
Quantity 25 g
UN Number 2226
Formula Weight 195.47
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Benzotrichloride
EINECSNumber (3)-0087
RTECSNumber XT9275000
TSCA Yes