missing translation for 'onlineSavingsMsg'
Learn More

Benzothiazole 96.0+%, TCI America™

Catalog No. B009225ML
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B009225ML 25 mL
B0092100ML 100 mL
B0092500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B009225ML Supplier TCI America Supplier No. B009225ML

Chemical Identifiers

CAS 95-16-9
Molecular Formula C7H5NS
Molecular Weight (g/mol) 135.184
MDL Number MFCD00005775
InChI Key IOJUPLGTWVMSFF-UHFFFAOYSA-N
Synonym benzothiazole, benzo d thiazole, benzosulfonazole, 1-thia-3-azaindene, vangard bt, benzothiazol, usaf ek-4812, benzthiazole, fema number 3256, unii-g5bw2593ep
PubChem CID 7222
ChEBI CHEBI:45993
IUPAC Name 1,3-benzothiazole
SMILES C1=CC=C2C(=C1)N=CS2

Specifications

Melting Point 2°C
Color Yellow
Boiling Point 231°C
Quantity 25 mL
UN Number 2810
Formula Weight 135.18
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material Benzothiazole
EINECSNumber (5)-3426
RTECSNumber DL0875000
TSCA Yes