missing translation for 'onlineSavingsMsg'
Learn More

Benzothiazole-6-carboxylic acid, 96%

Catalog No. p-7030964
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3186814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH3186814 Supplier Thermo Scientific Chemicals Supplier No. H3186814

CAS: 3622-35-3 | C8H5NO2S | 179.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3622-35-3
Molecular Formula C8H5NO2S
Molecular Weight (g/mol) 179.19
MDL Number MFCD00111651
InChI Key DMPZJACLHDWUFS-UHFFFAOYSA-N
Synonym benzothiazole-6-carboxylic acid, 6-benzothiazolecarboxylic acid, benzo d thiazole-6-carboxylic acid, zlchem 73, pubchem9871, enamine_005417, benzothiazole-6-carboxylic, 6-benzothiazolecarboxylicacid, 6-benzothiazole carboxylic acid, cl-cob-iii-274-2
PubChem CID 601670
IUPAC Name 1,3-benzothiazole-6-carboxylic acid
SMILES OC(=O)C1=CC=C2N=CSC2=C1

Specifications

Melting Point 245°C to 251°C
Quantity 25 g
Formula Weight 179.2
Percent Purity 96%
Chemical Name or Material Benzothiazole-6-carboxylic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only