accessibility menu, dialog, popup

UserName

OOHSN

CAS RN 3622-35-3

IUPAC Name: 1,3-benzothiazole-6-carboxylic acid
Synonyms: 1,3-benzothiazole-6-carboxylic acid
Molecular Weight (g/mol): 179.19
Molecular Formula: C8H5NO2S
InChi Key: DMPZJACLHDWUFS-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=C2N=CSC2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results