missing translation for 'onlineSavingsMsg'
Learn More

Benzothiazole-2-carbonyl chloride, 95%

CAS: 67748-61-2 | C8H4ClNOS | 197.636 g/mol

$101.34 - $264.79

Chemical Identifiers

CAS 67748-61-2
Molecular Formula C8H4ClNOS
Molecular Weight (g/mol) 197.636
MDL Number MFCD03659697
InChI Key AOIGQLLPWDXVGB-UHFFFAOYSA-N
Synonym benzothiazole-2-carbonyl chloride, 2-benzothiazolecarbonyl chloride, benzo d thiazole-2-carbonyl chloride, acmc-1b36m, 2-benzothiazolecarbonylchloride, benzothiazol-2-carbonyl chloride, benzothiazole-2-carbonyl chloride ., 2-benzothiazolecarbonyl chloride 9ci, benzothiazole-2-carboxylic acid chloride
PubChem CID 2776254
IUPAC Name 1,3-benzothiazole-2-carbonyl chloride
SMILES C1=CC=C2C(=C1)N=C(S2)C(=O)Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL19631MD
View Documents
Thermo Scientific Chemicals
L19631MD
250 mg
Each for $101.34
Only null left
 
AAL1963103
View Documents
Thermo Scientific Chemicals
L1963103
1 g
Each for $264.79
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

67748-61-2
197.636
AOIGQLLPWDXVGB-UHFFFAOYSA-N
2776254
C1=CC=C2C(=C1)N=C(S2)C(=O)Cl
C8H4ClNOS
MFCD03659697
benzothiazole-2-carbonyl chloride, 2-benzothiazolecarbonyl chloride, benzo d thiazole-2-carbonyl chloride, acmc-1b36m, 2-benzothiazolecarbonylchloride, benzothiazol-2-carbonyl chloride, benzothiazole-2-carbonyl chloride ., 2-benzothiazolecarbonyl chloride 9ci, benzothiazole-2-carboxylic acid chloride
1,3-benzothiazole-2-carbonyl chloride

Specifications

67748-61-2
C8H4ClNOS
250 mg
130990
benzothiazole-2-carbonyl chloride, 2-benzothiazolecarbonyl chloride, benzo d thiazole-2-carbonyl chloride, acmc-1b36m, 2-benzothiazolecarbonylchloride, benzothiazol-2-carbonyl chloride, benzothiazole-2-carbonyl chloride ., 2-benzothiazolecarbonyl chloride 9ci, benzothiazole-2-carboxylic acid chloride
C1=CC=C2C(=C1)N=C(S2)C(=O)Cl
197.636
197.64
Benzothiazole-2-carbonyl chloride
113°C to 115°C
MFCD03659697
UN3261
Moisture sensitive
AOIGQLLPWDXVGB-UHFFFAOYSA-N
1,3-benzothiazole-2-carbonyl chloride
2776254
95%
Videos
Safety and Handling

Safety and Handling

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

EINECSNumber : 000-000-0

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only