accessibility menu, dialog, popup

UserName

CAS RN 67748-61-2

IUPAC Name: 1,3-benzothiazole-2-carbonyl chloride
Synonyms: 1,3-benzothiazole-2-carbonyl chloride
Molecular Weight (g/mol): 197.64
Molecular Formula: C8H4ClNOS
InChi Key: AOIGQLLPWDXVGB-UHFFFAOYSA-N
SMILES: ClC(=O)C1=NC2=CC=CC=C2S1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results