missing translation for 'onlineSavingsMsg'
Learn More

Benzothiazole-2-carbonyl chloride, 95%

Catalog No. AAL19631MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL19631MD 250 mg
AAL1963103 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL19631MD Supplier Thermo Scientific Chemicals Supplier No. L19631MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 67748-61-2 | C8H4ClNOS | 197.636 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 67748-61-2
Molecular Formula C8H4ClNOS
Molecular Weight (g/mol) 197.636
MDL Number MFCD03659697
InChI Key AOIGQLLPWDXVGB-UHFFFAOYSA-N
Synonym benzothiazole-2-carbonyl chloride, 2-benzothiazolecarbonyl chloride, benzo d thiazole-2-carbonyl chloride, acmc-1b36m, 2-benzothiazolecarbonylchloride, benzothiazol-2-carbonyl chloride, benzothiazole-2-carbonyl chloride ., 2-benzothiazolecarbonyl chloride 9ci, benzothiazole-2-carboxylic acid chloride
PubChem CID 2776254
IUPAC Name 1,3-benzothiazole-2-carbonyl chloride
SMILES C1=CC=C2C(=C1)N=C(S2)C(=O)Cl

Specifications

Melting Point 113°C to 115°C
Quantity 250 mg
UN Number UN3261
Beilstein 130990
Sensitivity Moisture sensitive
Formula Weight 197.64
Percent Purity 95%
Chemical Name or Material Benzothiazole-2-carbonyl chloride
Hazard Category H314-H335
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only