missing translation for 'onlineSavingsMsg'
Learn More

(-)-Benzotetramisole 98.0+%, TCI America™

Catalog No. B35491G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B35491G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B35491G Supplier TCI America Supplier No. B35491G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 950194-37-3
Molecular Formula C15H12N2S
Molecular Weight (g/mol) 252.335
InChI Key YGCWPCVAVSIFLO-GFCCVEGCSA-N
Synonym (-)-BTM, (2S)-2,3-Dihydro-2-phenylimidazo[2,1-b]benzothiazole
PubChem CID 17747785
IUPAC Name (2S)-2-phenyl-1,2-dihydroimidazo[2,1-b][1,3]benzothiazole
SMILES C1C(N=C2N1C3=CC=CC=C3S2)C4=CC=CC=C4

Specifications

Melting Point 95°C
Color White-Yellow
Quantity 1 g
Formula Weight 252.34
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material (-)-Benzotetramisole