missing translation for 'onlineSavingsMsg'
Learn More

Benzopinacole, 98%, Thermo Scientific™

$306.56

Chemical Identifiers

CAS 464-72-2
Molecular Formula C26H22O2
Molecular Weight (g/mol) 366.45
MDL Number MFCD00004448
InChI Key MFEWNFVBWPABCX-UHFFFAOYSA-N
Synonym benzopinacol, benzopinacole, benzopinacone, benzpinacol, benzpinacone, tetraphenylethylene glycol, tetraphenyl-1,2-ethanediol, benzophenone pinacol, 1,1,2,2-tetraphenyl-1,2-ethanediol, 1,2-ethanediol, 1,1,2,2-tetraphenyl
PubChem CID 94766
IUPAC Name 1,1,2,2-tetraphenylethane-1,2-diol
SMILES C1=CC=C(C=C1)C(C2=CC=CC=C2)(C(C3=CC=CC=C3)(C4=CC=CC=C4)O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
AC105590100
View Documents Promotion Details
N/A 10g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC105591000
View Documents Promotion Details
N/A 100g Plastic bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

464-72-2
366.45
MFEWNFVBWPABCX-UHFFFAOYSA-N
94766
C1=CC=C(C=C1)C(C2=CC=CC=C2)(C(C3=CC=CC=C3)(C4=CC=CC=C4)O)O
C26H22O2
MFCD00004448
benzopinacol, benzopinacole, benzopinacone, benzpinacol, benzpinacone, tetraphenylethylene glycol, tetraphenyl-1,2-ethanediol, benzophenone pinacol, 1,1,2,2-tetraphenyl-1,2-ethanediol, 1,2-ethanediol, 1,1,2,2-tetraphenyl
1,1,2,2-tetraphenylethane-1,2-diol

Specifications

464-72-2
100.0
97.5% min. (HPLC)
(C6H5)2C(OH)C(OH)(C6H5)2
06, 1058
benzopinacol, benzopinacole, benzopinacone, benzpinacol, benzpinacone, tetraphenylethylene glycol, tetraphenyl-1,2-ethanediol, benzophenone pinacol, 1,1,2,2-tetraphenyl-1,2-ethanediol, 1,2-ethanediol, 1,1,2,2-tetraphenyl
MFEWNFVBWPABCX-UHFFFAOYSA-N
1,1,2,2-tetraphenylethane-1,2-diol
94766
98%
97.5
Authentic
C26H22O2
MFCD00004448
15, 1103
Solubility in water: insoluble
C1=CC=C(C=C1)C(C2=CC=CC=C2)(C(C3=CC=CC=C3)(C4=CC=CC=C4)O)O
366.45
366.45
Benzopinacole, 98%
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 207-356-8

RTECSNumber : KI2750000

TSCA : TSCA

SDS
Documents

Documents

RUO – Research Use Only