missing translation for 'onlineSavingsMsg'
Learn More

Benzopinacol 95.0+%, TCI America™

Supplier:  TCI America B103725G

 View more versions of this product

Catalog No. B103725G

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Benzopinacol
181°C
C26H22O2
25 g
benzopinacol, benzopinacole, benzopinacone, benzpinacol, benzpinacone, tetraphenylethylene glycol, tetraphenyl-1,2-ethanediol, benzophenone pinacol, 1,1,2,2-tetraphenyl-1,2-ethanediol, 1,2-ethanediol, 1,1,2,2-tetraphenyl
C1=CC=C(C=C1)C(C2=CC=CC=C2)(C(C3=CC=CC=C3)(C4=CC=CC=C4)O)O
366.46
366.46
Crystalline Powder
464-72-2
White
MFCD00004448
3077
MFEWNFVBWPABCX-UHFFFAOYSA-N
1,1,2,2-tetraphenylethane-1,2-diol
94766
≥95.0% (HPLC)

Chemical Identifiers

464-72-2
366.46
MFEWNFVBWPABCX-UHFFFAOYSA-N
94766
C1=CC=C(C=C1)C(C2=CC=CC=C2)(C(C3=CC=CC=C3)(C4=CC=CC=C4)O)O
C26H22O2
MFCD00004448
benzopinacol, benzopinacole, benzopinacone, benzpinacol, benzpinacone, tetraphenylethylene glycol, tetraphenyl-1,2-ethanediol, benzophenone pinacol, 1,1,2,2-tetraphenyl-1,2-ethanediol, 1,2-ethanediol, 1,1,2,2-tetraphenyl
1,1,2,2-tetraphenylethane-1,2-diol
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : KI2750000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions