missing translation for 'onlineSavingsMsg'
Learn More

Benzophenone-2,4'-dicarboxylic Acid Monohydrate 98.0+%, TCI America™

Catalog No. b008425g
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B008425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B008425G Supplier TCI America Supplier No. B008425G
Only null left

Chemical Identifiers

CAS 85-58-5
Molecular Formula C15H10O5
Molecular Weight (g/mol) 270.24
MDL Number MFCD04038016
InChI Key RWHRIIMYBNGFEV-UHFFFAOYSA-N
Synonym 2,4′C-Carbonyldibenzoic Acid
PubChem CID 221116
IUPAC Name 2-(4-carboxybenzoyl)benzoic acid
SMILES C1=CC=C(C(=C1)C(=O)C2=CC=C(C=C2)C(=O)O)C(=O)O

Specifications

Color White
Quantity 25 g
Formula Weight 270.24
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Benzophenone-2,4′-dicarboxylic Acid Monohydrate
EINECSNumber (4)-0134
TSCA No