accessibility menu, dialog, popup

UserName

HOOOHOO

CAS RN 85-58-5

IUPAC Name: 4-(2-carboxybenzoyl)benzoic acid
Synonyms: 4-(2-carboxybenzoyl)benzoic acid
Molecular Weight (g/mol): 270.24
Molecular Formula: C15H10O5
InChi Key: RWHRIIMYBNGFEV-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=C(C=C1)C(=O)C1=CC=CC=C1C(O)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results