missing translation for 'onlineSavingsMsg'
Learn More

Benzophenone-2,4,5-tricarboxylic Acid 98.0+%, TCI America™

Catalog No. B24211G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B24211G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B24211G Supplier TCI America Supplier No. B24211G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 135989-69-4
Molecular Formula C16H10O7
Molecular Weight (g/mol) 314.249
InChI Key OGQPSIOWWYARAZ-UHFFFAOYSA-N
Synonym 5-Benzoylbenzene-1,2,4-tricarboxylic Acid
PubChem CID 11522523
IUPAC Name 5-benzoylbenzene-1,2,4-tricarboxylic acid
SMILES C1=CC=C(C=C1)C(=O)C2=CC(=C(C=C2C(=O)O)C(=O)O)C(=O)O

Specifications

Color White-Yellow
Quantity 1 g
Formula Weight 314.25
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Benzophenone-2,4,5-tricarboxylic Acid