missing translation for 'onlineSavingsMsg'
Learn More

Benzonitrile, 99%, pure

CAS: 100-47-0 | C7H5N | 103.12 g/mol

Supplier:  Thermo Scientific Chemicals 105540025

 View more versions of this product

Catalog No. AC105540025


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Benzonitrile
98.5
-13.0°C
1.0100g/mL
71°C
98.5% min. (GC)
C7H5N
C6H5CN
2.5 L
1.01
phenyl cyanide, cyanobenzene, benzenenitrile, benzoic acid nitrile, benzene, cyano, benzenecarbonitrile, phenylcyanide, fenylkyanid, fenylkyanid czech, benzonitril
JFDZBHWFFUWGJE-UHFFFAOYSA-N
benzonitrile
7505
CHEBI:27991
99%
Liquid
100-47-0
100.0
Colorless to Yellow
188.0°C to 191.0°C
Authentic
Glass bottle
1.5270 to 1.5290
MFCD00001770
09, 275
15, 1099
Solubility in water: 10g/L (100°C). Other solubilities: misicble with most common organic solvents
C1=CC=C(C=C1)C#N
103.12
1.24 mPa.s (25°C)
103.12
Pure

Chemical Identifiers

100-47-0
JFDZBHWFFUWGJE-UHFFFAOYSA-N
7505
benzonitrile
MFCD00001770
phenyl cyanide, cyanobenzene, benzenenitrile, benzoic acid nitrile, benzene, cyano, benzenecarbonitrile, phenylcyanide, fenylkyanid, fenylkyanid czech, benzonitril
CHEBI:27991
C1=CC=C(C=C1)C#N
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful in contact with skin.
Harmful if swallowed.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

GHS Signal Word: Warning

EINECSNumber : 202-855-7

Recommended Storage : May darken during storage

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only