missing translation for 'onlineSavingsMsg'
Learn More

Benzonitrile, 99%, pure

Catalog No. p-3735574
Change view
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC105540010 1 L Glass bottle
AC105540025 2.5 L Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC105540010 Supplier Thermo Scientific Chemicals Supplier No. 105540010
Only null left

CAS: 100-47-0 | C7H5N | 103.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 100-47-0
MDL Number MFCD00001770
InChI Key JFDZBHWFFUWGJE-UHFFFAOYSA-N
Synonym phenyl cyanide, cyanobenzene, benzenenitrile, benzoic acid nitrile, benzene, cyano, benzenecarbonitrile, phenylcyanide, fenylkyanid, fenylkyanid czech, benzonitril
PubChem CID 7505
ChEBI CHEBI:27991
IUPAC Name benzonitrile
SMILES C1=CC=C(C=C1)C#N

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -13.0°C
Color Colorless to Yellow
Density 1.0100g/mL
Boiling Point 188.0°C to 191.0°C
Flash Point 71°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Molecular Formula C7H5N
Refractive Index 1.5270 to 1.5290
Linear Formula C6H5CN
Quantity 1 L
Beilstein 09, 275
Specific Gravity 1.01
Merck Index 15, 1099
Solubility Information Solubility in water: 10g/L (100°C). Other solubilities: misicble with most common organic solvents
Molecular Weight (g/mol) 103.12
Viscosity 1.24 mPa.s (25°C)
Formula Weight 103.12
Percent Purity 99%
Grade Pure
Physical Form Liquid
Chemical Name or Material Benzonitrile
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful in contact with skin.
Harmful if swallowed.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
EINECSNumber 202-855-7
Recommended Storage May darken during storage

RUO – Research Use Only