missing translation for 'onlineSavingsMsg'
Learn More

Benzoin Isopropyl Ether 99.0+%, TCI America™

Catalog No. B0942500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B0942500G 500 g
B094225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B0942500G Supplier TCI America Supplier No. B0942500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 6652-28-4
Molecular Formula C17H18O2
Molecular Weight (g/mol) 254.33
MDL Number MFCD00015025
InChI Key MSAHTMIQULFMRG-UHFFFAOYNA-N
Synonym benzoin isopropyl ether, 2-isopropoxy-1,2-diphenylethanone, ethanone, 2-1-methylethoxy-1,2-diphenyl, 2-isopropoxy-2-phenylacetophenone, benzoinisopropylether, acetophenone, 2-isopropoxy-2-phenyl, cbmicro_021426, 2-1-methylethoxy-1,2-diphenylethan-1-one, cambridge id 5476007
PubChem CID 110912
IUPAC Name 1,2-diphenyl-2-(propan-2-yloxy)ethan-1-one
SMILES CC(C)OC(C(=O)C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 79°C
Color White-Yellow
Boiling Point 180°C
Quantity 500 g
Formula Weight 254.33
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Benzoin Isopropyl Ether
EINECSNumber (4)-0079
TSCA Yes