accessibility menu, dialog, popup

UserName

CAS RN 6652-28-4

IUPAC Name: 1,2-diphenyl-2-(propan-2-yloxy)ethan-1-one
Synonyms: 2-isopropoxy-1,2-diphenylethanone
Molecular Weight (g/mol): 254.33
Molecular Formula: C17H18O2
InChi Key: MSAHTMIQULFMRG-UHFFFAOYNA-N
SMILES: CC(C)OC(C(=O)C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results