missing translation for 'onlineSavingsMsg'
Learn More

Benzoin Isobutyl Ether 94.0+%, TCI America™

Catalog No. p-7118805
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B101525G 25 g
B1015500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B101525G Supplier TCI America Supplier No. B101525G

Chemical Identifiers

CAS 22499-12-3
Molecular Formula C18H20O2
Molecular Weight (g/mol) 268.356
MDL Number MFCD00008933
InChI Key JMVZGKVGQDHWOI-UHFFFAOYSA-N
Synonym 2-Isobutoxy-2-phenylacetophenone
PubChem CID 90794
IUPAC Name 2-(2-methylpropoxy)-1,2-diphenylethanone
SMILES CC(C)COC(C1=CC=CC=C1)C(=O)C2=CC=CC=C2

Specifications

Boiling Point 133°C
Quantity 25 g
Formula Weight 268.36
Percent Purity ≥94.0% (GC)
Physical Form Liquid
Chemical Name or Material Benzoin Isobutyl Ether
EINECSNumber (4)-0079
RTECSNumber KM5782265
TSCA Yes