missing translation for 'onlineSavingsMsg'
Learn More

Benzoic Acid Zone Refined (number of passes:20) 99.0+%, TCI America™

Catalog No. B0062ZONE
Change view
Click to view available options
Quantity:
ZONE
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B0062ZONE ZONE
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B0062ZONE Supplier TCI America Supplier No. B00621SAMPLE

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 65-85-0
Molecular Formula C7H6O2
Molecular Weight (g/mol) 122.123
MDL Number MFCD00002398
InChI Key WPYMKLBDIGXBTP-UHFFFAOYSA-N
Synonym benzenecarboxylic acid, dracylic acid, benzeneformic acid, carboxybenzene, phenylformic acid, benzenemethanoic acid, phenylcarboxylic acid, retardex, benzoesaeure gk, benzoesaeure gv
PubChem CID 243
ChEBI CHEBI:30746
IUPAC Name benzoic acid
SMILES C1=CC=C(C=C1)C(=O)O

Specifications

Melting Point 125°C
Color White
pH 2.8 (saturated soln. 25°C)
Boiling Point 249°C
Quantity ZONE
Formula Weight 122.12
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Benzoic Acid Zone Refined (number of passes:20)
EINECSNumber (3)-1397
RTECSNumber DG0875000
TSCA Yes