missing translation for 'onlineSavingsMsg'
Learn More

Medisca Benzoic Acid USP

Catalog No. 14185644
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14185644 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14185644 Supplier Medisca Supplier No. 061505
Get 1 Extra Chemical When You Buy 4
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Benzoic Acid USP ranges from white crystals, scales, or needles. Benzoic Acid has a slight odor, usually suggesting benzaldehyde or benzoin.

Benzoic Acid USP ranges from white crystals, scales, or needles. Benzoic Acid has a slight odor, usually suggesting benzaldehyde or benzoin.

Specifications

Chemical Name or Material Benzoic Acid USP
Packaging Jar, 300mL, White, HDPE, Round, 63-400, TE Pano Neck
CAS 65-85-0
Type Drug Standard
Molecular Weight (g/mol) 122.12
Quantity 100 g
Percent Purity 99.5 to 100.5 %
Grade USP