missing translation for 'onlineSavingsMsg'
Learn More

Benzoic Acid 99.0+%, TCI America™

Catalog No. B263525G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B263525G 25 g
B2635500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B263525G Supplier TCI America Supplier No. B263525G

Chemical Identifiers

CAS 65-85-0
Molecular Formula C7H6O2
Molecular Weight (g/mol) 122.123
MDL Number MFCD00002398
InChI Key WPYMKLBDIGXBTP-UHFFFAOYSA-N
Synonym benzenecarboxylic acid, dracylic acid, benzeneformic acid, carboxybenzene, phenylformic acid, benzenemethanoic acid, phenylcarboxylic acid, retardex, benzoesaeure gk, benzoesaeure gv
PubChem CID 243
ChEBI CHEBI:30746
IUPAC Name benzoic acid
SMILES C1=CC=C(C=C1)C(=O)O

Specifications

Melting Point 124°C
Color White
Boiling Point 249°C
Quantity 25 g
Formula Weight 122.12
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Benzoic Acid
EINECSNumber (3)-1397
RTECSNumber DG0875000
TSCA Yes