missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzoic Acid, Crystal, Reagent, ACS, 99.5%, Spectrum™ Chemical
B1085, 65-85-0, C7H6O2
Supplier: Spectrum Chemical Mfg Cor B1085500GM
Description
Spectrum™ Chemical Benzoic Acid, Crystal, Reagent, ACS an aromatic crystalline solid that is colorless. It is used as a standard for calibrating bomb calorimeters in teaching laboratories and as a synthetic precursor to a series of plasticizers. As an ACS grade quality reagent, its chemical specifications are the de facto standards for chemicals used in many high-purity applications and typically designate the highest quality chemical available for laboratory use. Spectrum Chemical manufactured Reagent ACS grade products meet the toughest regulatory standards for quality and purity.Specifications
| 65-85-0,65-85-0 | |
| Poly Bottle | |
| 500 g | |
| OC(=O)C1=CC=CC=C1 | |
| 122.12 | |
| ACS |
| 100%,100% | |
| C7H6O2 | |
| WPYMKLBDIGXBTP-UHFFFAOYSA-N | |
| benzoic acid | |
| 99.5% | |
| Crystals or Flakes |
Chemical Identifiers
| 65-85-0 | |
| 122.12 | |
| benzoic acid |
| C7H6O2 | |
| WPYMKLBDIGXBTP-UHFFFAOYSA-N | |
| OC(=O)C1=CC=CC=C1 |