missing translation for 'onlineSavingsMsg'
Learn More

Benzoic Acid, Crystal, Reagent, ACS, 99.5%, Spectrum™ Chemical

Catalog No. 18600897
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Poly Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18600897 125 g Poly Bottle
18600898 500 g Poly Bottle
18600899 2.5 kg Poly Bottle
18600900 12 kg Poly Bottle
18600901 50 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18600897 Supplier Spectrum Chemical Mfg Cor Supplier No. B1085125GM

B1085, 65-85-0, C7H6O2

Spectrum™ Chemical Benzoic Acid, Crystal, Reagent, ACS an aromatic crystalline solid that is colorless. It is used as a standard for calibrating bomb calorimeters in teaching laboratories and as a synthetic precursor to a series of plasticizers. As an ACS grade quality reagent, its chemical specifications are the de facto standards for chemicals used in many high-purity applications and typically designate the highest quality chemical available for laboratory use. Spectrum Chemical manufactured Reagent ACS grade products meet the toughest regulatory standards for quality and purity.

Chemical Identifiers

CAS 65-85-0
Molecular Formula C7H6O2
Molecular Weight (g/mol) 122.12
InChI Key WPYMKLBDIGXBTP-UHFFFAOYSA-N
IUPAC Name benzoic acid
SMILES OC(=O)C1=CC=CC=C1

Specifications

CAS Max % 100%
Packaging Poly Bottle
Quantity 125 g
Percent Purity 99.5%
Grade ACS
Physical Form Crystals or Flakes