Learn More
Benzoic acid, 99.6%, ACS reagent
CAS: 65-85-0 | C7H6O2 | 122.12 g/mol
Supplier: Thermo Scientific Chemicals 423470020
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Benzoic acid | |
| 99.5 | |
| 121.0°C to 123.0°C | |
| 249.0°C | |
| 0.005% max. | |
| 99.6% | |
| C7H6O2 | |
| 2 kg | |
| 01,49; 13,312 | |
| benzenecarboxylic acid, dracylic acid, benzeneformic acid, carboxybenzene, phenylformic acid, benzenemethanoic acid, phenylcarboxylic acid, retardex, benzoesaeure gk, benzoesaeure gv | |
| WPYMKLBDIGXBTP-UHFFFAOYSA-N | |
| benzoic acid | |
| 243 | |
| 122.12 | |
| ACS Reagent | |
| Crystalline Powder |
| 65-85-0 | |
| 100.0 | |
| White | |
| 121°C | |
| Authentic | |
| Plastic bottle | |
| C6H5CO2H | |
| 09,92 | |
| 15,1093 | |
| Solubility in water: 3g/L (20°C). Other solubilities: soluble in alcohol,benzene,ether,chloroform,,ccl4,carbon disulfide and turpentine | |
| C1=CC=C(C=C1)C(=O)O | |
| 122.12 | |
| CHEBI:30746 | |
| 99.6% | |
| 0.005% max. (in methanol) |
Chemical Identifiers
| 65-85-0 | |
| 122.12 | |
| benzenecarboxylic acid, dracylic acid, benzeneformic acid, carboxybenzene, phenylformic acid, benzenemethanoic acid, phenylcarboxylic acid, retardex, benzoesaeure gk, benzoesaeure gv | |
| CHEBI:30746 | |
| C1=CC=C(C=C1)C(=O)O |
| C7H6O2 | |
| WPYMKLBDIGXBTP-UHFFFAOYSA-N | |
| 243 | |
| benzoic acid |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye damage.
Causes damage to organs through prolonged or repeated exposure if inhaled.
GHS P Statement
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for s
GHS Signal Word: Danger
EINECSNumber : 200-618-2
RUO – Research Use Only