missing translation for 'onlineSavingsMsg'
Learn More

Benzohydroxamic acid, 98%

CAS: 495-18-1 | C7H7NO2 | 137.138 g/mol

$159.23 - $2015.97

Chemical Identifiers

CAS 495-18-1
Molecular Formula C7H7NO2
Molecular Weight (g/mol) 137.138
MDL Number MFCD00002109
InChI Key VDEUYMSGMPQMIK-UHFFFAOYSA-N
Synonym benzohydroxamic acid, benzhydroxamic acid, benzamide, n-hydroxy, benzohydroxamate, phenylhydroxamic acid, benzohyroxamate, hydroxylamine, n-benzoyl, benzoylhydroxamic acid, n-hydroxy-benzamide, benzenecarbohydroxamic acid
PubChem CID 10313
IUPAC Name N-hydroxybenzamide
SMILES C1=CC=C(C=C1)C(=O)NO
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1050909
View Documents
Thermo Scientific Chemicals
A1050909
10 g
Each for $159.23
Only null left
 
AAA1050914
View Documents
Thermo Scientific Chemicals
A1050914
25 g
Each for $330.11
Only null left
 
AAA1050918
View Documents
Thermo Scientific Chemicals
A1050918
50 g
Each for $499.58
Only null left
 
AAA1050930
View Documents
Thermo Scientific Chemicals
A1050930
250 g
Each for $2,015.97
Only null left
 
Description

Description

Benzhydroxamic acid is used as precursor in the synthesis of novel mono-anionic and di-anionic hydroxamato complexes by reacting with BiPh3 and Bi(O(t)Bu)3, which has anti-bacterial activity against helicobacter pylori. It is used in the photometric determination of trace amounts of vanadium in alloy steels by making mixed-ligand vanadium chelates with ammonium thiocyanate. It is also involved in the palladium catalyzed synthesis of benzisoxazolones.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzhydroxamic acid is used as precursor in the synthesis of novel mono-anionic and di-anionic hydroxamato complexes by reacting with BiPh3 and Bi(O(t)Bu)3, which has anti-bacterial activity against helicobacter pylori. It is used in the photometric determination of trace amounts of vanadium in alloy steels by making mixed-ligand vanadium chelates with ammonium thiocyanate. It is also involved in the palladium catalyzed synthesis of benzisoxazolones.

Solubility
Soluble in alcohol. Slightly soluble in ether. Insoluble in benzene.

Notes
Store in cool place. Incompatible with strong oxidizing agents.
Specifications

Chemical Identifiers

495-18-1
137.138
VDEUYMSGMPQMIK-UHFFFAOYSA-N
10313
C1=CC=C(C=C1)C(=O)NO
C7H7NO2
MFCD00002109
benzohydroxamic acid, benzhydroxamic acid, benzamide, n-hydroxy, benzohydroxamate, phenylhydroxamic acid, benzohyroxamate, hydroxylamine, n-benzoyl, benzoylhydroxamic acid, n-hydroxy-benzamide, benzenecarbohydroxamic acid
N-hydroxybenzamide

Specifications

495-18-1
C7H7NO2
10 g
benzohydroxamic acid, benzhydroxamic acid, benzamide, n-hydroxy, benzohydroxamate, phenylhydroxamic acid, benzohyroxamate, hydroxylamine, n-benzoyl, benzoylhydroxamic acid, n-hydroxy-benzamide, benzenecarbohydroxamic acid
VDEUYMSGMPQMIK-UHFFFAOYSA-N
N-hydroxybenzamide
10313
98%
∼125°C (decomposition)
MFCD00002109
1907585
Soluble in alcohol. Slightly soluble in ether. Insoluble in benzene.
C1=CC=C(C=C1)C(=O)NO
137.138
137.14
Benzohydroxamic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H341-H315-H319-H335
Suspected of causing genetic defects.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P201-P202-P261-P264b-P271-P280i-P281-P302+P352-P304+P340-P305+P351+P338-P308+P313-P332+P313-P362-P501c

H315-H319-H335-H341

EINECSNumber : 207-797-6

RTECSNumber : DF9650000

TSCA : Yes

Recommended Storage : Keep cold

SDS
Documents

Documents

RUO – Research Use Only