missing translation for 'onlineSavingsMsg'
Learn More

Benzohydroxamic acid, 98%

CAS: 495-18-1 | C7H7NO2 | 137.138 g/mol

Supplier:  Thermo Scientific Chemicals A1050918

 View more versions of this product

Catalog No. AAA1050918

Only null left
Explore available promotions

Description

Description

Benzhydroxamic acid is used as precursor in the synthesis of novel mono-anionic and di-anionic hydroxamato complexes by reacting with BiPh3 and Bi(O(t)Bu)3, which has anti-bacterial activity against helicobacter pylori. It is used in the photometric determination of trace amounts of vanadium in alloy steels by making mixed-ligand vanadium chelates with ammonium thiocyanate. It is also involved in the palladium catalyzed synthesis of benzisoxazolones.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzhydroxamic acid is used as precursor in the synthesis of novel mono-anionic and di-anionic hydroxamato complexes by reacting with BiPh3 and Bi(O(t)Bu)3, which has anti-bacterial activity against helicobacter pylori. It is used in the photometric determination of trace amounts of vanadium in alloy steels by making mixed-ligand vanadium chelates with ammonium thiocyanate. It is also involved in the palladium catalyzed synthesis of benzisoxazolones.

Solubility
Soluble in alcohol. Slightly soluble in ether. Insoluble in benzene.

Notes
Store in cool place. Incompatible with strong oxidizing agents.
Specifications

Specifications

Benzohydroxamic acid
∼125°C (decomposition)
MFCD00002109
1907585
Soluble in alcohol. Slightly soluble in ether. Insoluble in benzene.
C1=CC=C(C=C1)C(=O)NO
137.138
137.14
495-18-1
C7H7NO2
50 g
benzohydroxamic acid, benzhydroxamic acid, benzamide, n-hydroxy, benzohydroxamate, phenylhydroxamic acid, benzohyroxamate, hydroxylamine, n-benzoyl, benzoylhydroxamic acid, n-hydroxy-benzamide, benzenecarbohydroxamic acid
VDEUYMSGMPQMIK-UHFFFAOYSA-N
N-hydroxybenzamide
10313
98%

Chemical Identifiers

495-18-1
137.138
VDEUYMSGMPQMIK-UHFFFAOYSA-N
10313
C1=CC=C(C=C1)C(=O)NO
C7H7NO2
MFCD00002109
benzohydroxamic acid, benzhydroxamic acid, benzamide, n-hydroxy, benzohydroxamate, phenylhydroxamic acid, benzohyroxamate, hydroxylamine, n-benzoyl, benzoylhydroxamic acid, n-hydroxy-benzamide, benzenecarbohydroxamic acid
N-hydroxybenzamide
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H341-H315-H319-H335
Suspected of causing genetic defects.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P201-P202-P261-P264b-P271-P280i-P281-P302+P352-P304+P340-P305+P351+P338-P308+P313-P332+P313-P362-P501c

H315-H319-H335-H341

EINECSNumber : 207-797-6

RTECSNumber : DF9650000

TSCA : Yes

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only