missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzoguanamine 99.0+%, TCI America™
$39.14 - $93.21
Chemical Identifiers
| CAS | 91-76-9 |
|---|---|
| Molecular Formula | C9H9N5 |
| Molecular Weight (g/mol) | 187.21 |
| MDL Number | MFCD00023187 |
| InChI Key | GZVHEAJQGPRDLQ-UHFFFAOYSA-N |
| Synonym | benzoguanamine, 2,4-diamino-6-phenyl-1,3,5-triazine, benzoguanimine, 1,3,5-triazine-2,4-diamine, 6-phenyl, usaf rh-5, 2,4-diamino-6-phenyl-s-triazine, 2-phenyl-4,6-diamino-s-triazine, 4,6-diamino-2-phenyl-s-triazine, benzoguanamine van, s-triazine, 2,4-diamino-6-phenyl |
| PubChem CID | 7064 |
| IUPAC Name | 6-phenyl-1,3,5-triazine-2,4-diamine |
| SMILES | NC1=NC(=NC(N)=N1)C1=CC=CC=C1 |
Chemical Identifiers
| 91-76-9 | |
| 187.21 | |
| GZVHEAJQGPRDLQ-UHFFFAOYSA-N | |
| 7064 | |
| NC1=NC(=NC(N)=N1)C1=CC=CC=C1 |
| C9H9N5 | |
| MFCD00023187 | |
| benzoguanamine, 2,4-diamino-6-phenyl-1,3,5-triazine, benzoguanimine, 1,3,5-triazine-2,4-diamine, 6-phenyl, usaf rh-5, 2,4-diamino-6-phenyl-s-triazine, 2-phenyl-4,6-diamino-s-triazine, 4,6-diamino-2-phenyl-s-triazine, benzoguanamine van, s-triazine, 2,4-diamino-6-phenyl | |
| 6-phenyl-1,3,5-triazine-2,4-diamine |
Specifications
| 91-76-9 | |
| White | |
| MFCD00023187 | |
| benzoguanamine, 2,4-diamino-6-phenyl-1,3,5-triazine, benzoguanimine, 1,3,5-triazine-2,4-diamine, 6-phenyl, usaf rh-5, 2,4-diamino-6-phenyl-s-triazine, 2-phenyl-4,6-diamino-s-triazine, 4,6-diamino-2-phenyl-s-triazine, benzoguanamine van, s-triazine, 2,4-diamino-6-phenyl | |
| NC1=NC(=NC(N)=N1)C1=CC=CC=C1 | |
| 187.21 | |
| 187.21 | |
| Crystalline Powder |
| 228°C | |
| C9H9N5 | |
| 25 g | |
| GZVHEAJQGPRDLQ-UHFFFAOYSA-N | |
| 6-phenyl-1,3,5-triazine-2,4-diamine | |
| 7064 | |
| ≥99.0% (HPLC) | |
| Benzoguanamine |
Safety and Handling
EINECSNumber : (5)-1028
RTECSNumber : XY7000000
TSCA : Yes