accessibility menu, dialog, popup

UserName

NH2NNH2NN

CAS RN 91-76-9

IUPAC Name: 6-phenyl-1,3,5-triazine-2,4-diamine
Synonyms: benzoguanamine
Molecular Weight (g/mol): 187.21
Molecular Formula: C9H9N5
InChi Key: GZVHEAJQGPRDLQ-UHFFFAOYSA-N
SMILES: NC1=NC(=NC(N)=N1)C1=CC=CC=C1
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)