missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzoguanamine, 99%
CAS: 91-76-9 | C9H9N5 | 187.21 g/mol
$41.32 - $397.19
Chemical Identifiers
| CAS | 91-76-9 |
|---|---|
| Molecular Formula | C9H9N5 |
| Molecular Weight (g/mol) | 187.21 |
| MDL Number | MFCD00023187 |
| InChI Key | GZVHEAJQGPRDLQ-UHFFFAOYSA-N |
| Synonym | benzoguanamine, 2,4-diamino-6-phenyl-1,3,5-triazine, benzoguanimine, 1,3,5-triazine-2,4-diamine, 6-phenyl, usaf rh-5, 2,4-diamino-6-phenyl-s-triazine, 2-phenyl-4,6-diamino-s-triazine, 4,6-diamino-2-phenyl-s-triazine, benzoguanamine van, s-triazine, 2,4-diamino-6-phenyl |
| PubChem CID | 7064 |
| IUPAC Name | 6-phenyl-1,3,5-triazine-2,4-diamine |
| SMILES | NC1=NC(=NC(N)=N1)C1=CC=CC=C1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAB2015722
|
Thermo Scientific Chemicals
B2015722 |
100 g |
Each for $41.32
|
|
|||||
|
AAB2015736
|
Thermo Scientific Chemicals
B2015736 |
500 g |
Each for $112.81
|
|
|||||
|
AAB201570E
|
Thermo Scientific Chemicals
B201570E |
2500 g |
Each for $397.19
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 91-76-9 | |
| 187.21 | |
| GZVHEAJQGPRDLQ-UHFFFAOYSA-N | |
| 7064 | |
| NC1=NC(=NC(N)=N1)C1=CC=CC=C1 |
| C9H9N5 | |
| MFCD00023187 | |
| benzoguanamine, 2,4-diamino-6-phenyl-1,3,5-triazine, benzoguanimine, 1,3,5-triazine-2,4-diamine, 6-phenyl, usaf rh-5, 2,4-diamino-6-phenyl-s-triazine, 2-phenyl-4,6-diamino-s-triazine, 4,6-diamino-2-phenyl-s-triazine, benzoguanamine van, s-triazine, 2,4-diamino-6-phenyl | |
| 6-phenyl-1,3,5-triazine-2,4-diamine |
Specifications
| 91-76-9 | |
| C9H9N5 | |
| 100 g | |
| 14,1089 | |
| GZVHEAJQGPRDLQ-UHFFFAOYSA-N | |
| 6-phenyl-1,3,5-triazine-2,4-diamine | |
| 7064 | |
| 99% |
| 225°C to 228°C | |
| MFCD00023187 | |
| 153223 | |
| benzoguanamine, 2,4-diamino-6-phenyl-1,3,5-triazine, benzoguanimine, 1,3,5-triazine-2,4-diamine, 6-phenyl, usaf rh-5, 2,4-diamino-6-phenyl-s-triazine, 2-phenyl-4,6-diamino-s-triazine, 4,6-diamino-2-phenyl-s-triazine, benzoguanamine van, s-triazine, 2,4-diamino-6-phenyl | |
| NC1=NC(=NC(N)=N1)C1=CC=CC=C1 | |
| 187.21 | |
| 187.21 | |
| Benzoguanamine |
Safety and Handling
GHS H Statement
H301
Toxic if swallowed.
P261-P264b-P270-P271-P301+P312-P304+P340-P312-P330-P501c
H302+H332
EINECSNumber : 202-095-6
RTECSNumber : XY7000000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only