missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzo[f]quinoline 98.0+%, TCI America™
Supplier: TCI America B00871G
Specifications
| Benzo[f]quinoline | |
| 93°C | |
| 350°C | |
| MFCD00022235 | |
| benzo f quinoline, 5,6-benzoquinoline, 1-azaphenanthrene, benzo f-quinoline, beta-naphthoquinoline, .beta.-naphthoquinoline, 5,6-benzo f quinoline, ccris 800, unii-525476dtml | |
| C1=CC=C2C(=C1)C=CC3=C2C=CC=N3 | |
| 179.222 | |
| 179.22 | |
| Crystalline Powder |
| 85-02-9 | |
| White-Yellow | |
| C13H9N | |
| 1 g | |
| HCAUQPZEWLULFJ-UHFFFAOYSA-N | |
| benzo[f]quinoline | |
| 6796 | |
| ≥98.0% (T) |
Chemical Identifiers
| 85-02-9 | |
| 179.222 | |
| HCAUQPZEWLULFJ-UHFFFAOYSA-N | |
| 6796 | |
| C1=CC=C2C(=C1)C=CC3=C2C=CC=N3 |
| C13H9N | |
| MFCD00022235 | |
| benzo f quinoline, 5,6-benzoquinoline, 1-azaphenanthrene, benzo f-quinoline, beta-naphthoquinoline, .beta.-naphthoquinoline, 5,6-benzo f quinoline, ccris 800, unii-525476dtml | |
| benzo[f]quinoline |
Safety and Handling
RTECSNumber : DK1428000
TSCA : No