accessibility menu, dialog, popup

UserName

CAS RN 85-02-9

N

CAS RN 85-02-9

IUPAC Name: benzo[f]quinoline
Synonyms: benzo(F)quinoline
Molecular Weight (g/mol): 179.22
Molecular Formula: C13H9N
InChi Key: HCAUQPZEWLULFJ-UHFFFAOYSA-N
SMILES: C1=CC=C2C(C=CC3=NC=CC=C23)=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results