missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzo[d]isoxazol-3-ol, 96%, Thermo Scientific™
Chemical Identifiers
| CAS | 21725-69-9 |
|---|---|
| Molecular Formula | C7H5NO2 |
| Molecular Weight (g/mol) | 135.12 |
| InChI Key | QLDQYRDCPNBPII-UHFFFAOYSA-N |
| Synonym | benzo d isoxazol-3-ol, 1,2-benzisoxazol-3 2h-one, 1,2-benzoxazol-3-ol, 1,2-benzisoxazol-3-ol, 3-hydroxybenzisoxazole, 3-hydroxy-1,2-benzisoxazole, 1,2-benzisoxazolin-3-one, 1,2-benzisoxazol-3-ol 8ci, 2h-1,2-benzoxazol-3-one, benzisoxazolone |
| PubChem CID | 210830 |
| IUPAC Name | 1,2-benzoxazol-3-one |
| SMILES | C1=CC=C2C(=C1)C(=O)NO2 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
AC450960010
|
N/A | 1g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
| 21725-69-9 | |
| 135.12 | |
| benzo d isoxazol-3-ol, 1,2-benzisoxazol-3 2h-one, 1,2-benzoxazol-3-ol, 1,2-benzisoxazol-3-ol, 3-hydroxybenzisoxazole, 3-hydroxy-1,2-benzisoxazole, 1,2-benzisoxazolin-3-one, 1,2-benzisoxazol-3-ol 8ci, 2h-1,2-benzoxazol-3-one, benzisoxazolone | |
| 1,2-benzoxazol-3-one |
| C7H5NO2 | |
| QLDQYRDCPNBPII-UHFFFAOYSA-N | |
| 210830 | |
| C1=CC=C2C(=C1)C(=O)NO2 |
Specifications
| 21725-69-9 | |
| Glass Bottle | |
| benzo d isoxazol-3-ol, 1,2-benzisoxazol-3 2h-one, 1,2-benzoxazol-3-ol, 1,2-benzisoxazol-3-ol, 3-hydroxybenzisoxazole, 3-hydroxy-1,2-benzisoxazole, 1,2-benzisoxazolin-3-one, 1,2-benzisoxazol-3-ol 8ci, 2h-1,2-benzoxazol-3-one, benzisoxazolone | |
| C1=CC=C2C(=C1)C(=O)NO2 | |
| 135.12 | |
| 135.12 | |
| Benzo[d]isoxazol-3-ol |
| 95% min. (HPLC) | |
| C7H5NO2 | |
| QLDQYRDCPNBPII-UHFFFAOYSA-N | |
| 1,2-benzoxazol-3-one | |
| 210830 | |
| 96% |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF
RUO – Research Use Only