missing translation for 'onlineSavingsMsg'
Learn More

Benzo[cd]indol-2(1H)-one 97.0+%, TCI America™

Catalog No. B302325G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B302325G 25 g
B30235G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B302325G Supplier TCI America Supplier No. B302325G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 130-00-7
Molecular Formula C11H7NO
Molecular Weight (g/mol) 169.183
MDL Number MFCD00009748
InChI Key GPYLCFQEKPUWLD-UHFFFAOYSA-N
Synonym Naphthostyril
PubChem CID 67222
IUPAC Name 1H-benzo[cd]indol-2-one
SMILES C1=CC2=C3C(=C1)C(=O)NC3=CC=C2

Specifications

Melting Point 181°C
Color Yellow
Quantity 25 g
Formula Weight 169.18
Percent Purity ≥97.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material Benzo[cd]indol-2(1H)-one
EINECSNumber (5)-0097
RTECSNumber DE3202000
TSCA Yes