accessibility menu, dialog, popup

UserName

CAS RN 130-00-7

IUPAC Name: 2-azatricyclo[6.3.1.0⁴,¹²]dodeca-1(12),4,6,8,10-pentaen-3-one
Synonyms: benz(cd)indol-2(1H)-one
Molecular Weight (g/mol): 169.18
Molecular Formula: C11H7NO
InChi Key: GPYLCFQEKPUWLD-UHFFFAOYSA-N
SMILES: O=C1NC2=C3C1=CC=CC3=CC=C2
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results