missing translation for 'onlineSavingsMsg'
Learn More

Benzo[b]thiophene-3-sulfonyl chloride, 97%

Catalog No. AAH61368MD
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH61368MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH61368MD Supplier Thermo Scientific Chemicals Supplier No. H61368MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 18494-87-6 | C8H5ClO2S2 | 232.696 g/mol

Benzo[b]thiophene-3-sulfonyl chloride is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzo[b]thiophene-3-sulfonyl chloride is used as a pharmaceutical intermediate.

Solubility
Sparingly soluble in water (0.091 g/L at 25°C).

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is sensitive to moisture.

Chemical Identifiers

CAS 18494-87-6
Molecular Formula C8H5ClO2S2
Molecular Weight (g/mol) 232.696
MDL Number MFCD02681897
InChI Key WYYRQMZTUXFUTC-UHFFFAOYSA-N
Synonym benzothiophene-3-sulfonyl chloride, benzo b thiophene-3-sulfonyl chloride, benzo b thiophene-3-sulfonylchloride, benzo b thiophene-3-sulphonyl chloride, pubchem14540, 3-chlorosulphonyl-benzothiophene, benzothiophene-3-sulfonylchloride, 3-chlorosulphonyl-1-benzothiophene, benzo b thiophen-3-ylchlorosulfone
PubChem CID 2776340
IUPAC Name 1-benzothiophene-3-sulfonyl chloride
SMILES C1=CC=C2C(=C1)C(=CS2)S(=O)(=O)Cl

Specifications

Melting Point 86°C to 88°C
Quantity 250 mg
UN Number UN3261
Beilstein 1285265
Sensitivity Moisture sensitive
Solubility Information Sparingly soluble in water (0.091g/L at 25°C).
Formula Weight 232.71
Percent Purity 97%
Chemical Name or Material Benzo[b]thiophene-3-sulfonyl chloride
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
TSCA No
Recommended Storage Keep cold; Store under Argon

RUO – Research Use Only