accessibility menu, dialog, popup

UserName

CAS RN 18494-87-6

SSOOCl

CAS RN 18494-87-6

IUPAC Name: 1-benzothiophene-3-sulfonyl chloride
Synonyms: 1-benzothiophene-3-sulfonyl chloride
Molecular Weight (g/mol): 232.7
Molecular Formula: C8H5ClO2S2
InChi Key: WYYRQMZTUXFUTC-UHFFFAOYSA-N
SMILES: ClS(=O)(=O)C1=CSC2=CC=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results