missing translation for 'onlineSavingsMsg'
Learn More

Benzo[b]thiophene-3-carboxylic Acid 98.0+%, TCI America™

Catalog No. p-7123668
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B4650200MG 200 mg
B46501G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B4650200MG Supplier TCI America Supplier No. B4650200MG
Only null left

Chemical Identifiers

CAS 5381-25-9
Molecular Formula C9H6O2S
Molecular Weight (g/mol) 178.205
MDL Number MFCD01846406
InChI Key DRBLTQNCQJXSNU-UHFFFAOYSA-N
Synonym benzo b thiophene-3-carboxylic acid, benzothiophene-3-carboxylic acid, benzothiophene-3-carboxylicacid, pubchem13463, acmc-20a10z, ksc178m8d, 1-benzo b thiophene-3-carboxylic acid, #, thianaphthene-3-carboxylic acid
PubChem CID 601280
IUPAC Name 1-benzothiophene-3-carboxylic acid
SMILES C1=CC=C2C(=C1)C(=CS2)C(=O)O

Specifications

Melting Point 180°C
Color Yellow
Quantity 200 mg
Formula Weight 178.21
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Benzo[b]thiophene-3-carboxylic Acid