accessibility menu, dialog, popup

UserName

SOOH

CAS RN 5381-25-9

IUPAC Name: 1-benzothiophene-3-carboxylic acid
Synonyms: 1-benzothiophene-3-carboxylic acid
Molecular Weight (g/mol): 178.21
Molecular Formula: C9H6O2S
InChi Key: DRBLTQNCQJXSNU-UHFFFAOYSA-N
SMILES: OC(=O)C1=CSC2=CC=CC=C12
Percent Purity 
  • (2)
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (5)
  • (2)
  • (3)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)