missing translation for 'onlineSavingsMsg'
Learn More

Benzo[b]thiophene-3-carboxaldehyde 98.0+%, TCI America™

Catalog No. p-7123339
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B40861G 1 g
B40865G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B40861G Supplier TCI America Supplier No. B40861G

Chemical Identifiers

CAS 5381-20-4
Molecular Formula C9H6OS
Molecular Weight (g/mol) 162.21
MDL Number MFCD00052376
InChI Key WDJLPQCBTBZTRH-UHFFFAOYSA-N
Synonym benzo b thiophene-3-carbaldehyde, benzo b thiophene-3-carboxaldehyde, thianaphthene-3-carboxaldehyde, 3-formylbenzo b thiophene, benzothiophene-3-carbaldehyde, 1-benzothiophen-3-carbaldehyd, pubchem13543, acmc-1apd4, thianapthene-3-carboxaldehyde, ksc274c6b
PubChem CID 227328
IUPAC Name 1-benzothiophene-3-carbaldehyde
SMILES O=CC1=CSC2=CC=CC=C12

Specifications

Melting Point 58°C
Color White-Yellow
Boiling Point 133°C
Quantity 1 g
Formula Weight 162.21
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Benzo[b]thiophene-3-carboxaldehyde