accessibility menu, dialog, popup

UserName

OS

CAS RN 5381-20-4

IUPAC Name: 1-benzothiophene-3-carbaldehyde
Synonyms: 1-benzothiophene-3-carbaldehyde
Molecular Weight (g/mol): 162.21
Molecular Formula: C9H6OS
InChi Key: WDJLPQCBTBZTRH-UHFFFAOYSA-N
SMILES: O=CC1=CSC2=CC=CC=C12
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results