missing translation for 'onlineSavingsMsg'
Learn More

Benzo[b]thiophene-3-boronic acid pinacol ester, 95%

CAS: 171364-86-6 | C14H17BO2S | 260.158 g/mol

$106.63 - $300.10

Chemical Identifiers

CAS 171364-86-6
Molecular Formula C14H17BO2S
Molecular Weight (g/mol) 260.158
MDL Number MFCD11110552
InChI Key LTSGSDOTQABJMA-UHFFFAOYSA-N
Synonym 2-benzo b thiophen-3-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-1-benzothiophen-3-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, benzo b thiophene-3-boronic acid pinacol ester, 2-benzo b thiophen-3-yl-4,4,5,5-tetramethyl-1,3,2 dioxaborolane, benzo b thiophene, 3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 2-benzo b thiophen-3-yl-4,4,5,5-tetramethyl-1,3,2-dioxaboro lane, 4,4,5,5-tetramethyl-2-benzo b thiophen-3-yl-1,3,2 dioxaborolane
PubChem CID 10658979
IUPAC Name 2-(1-benzothiophen-3-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CSC3=CC=CC=C23
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH51876MD
View Documents
Thermo Scientific Chemicals
H51876MD
250 mg
Each for $106.63
Only null left
 
AAH5187603
View Documents
Thermo Scientific Chemicals
H5187603
1 g
Each for $300.10
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

171364-86-6
260.158
LTSGSDOTQABJMA-UHFFFAOYSA-N
10658979
B1(OC(C(O1)(C)C)(C)C)C2=CSC3=CC=CC=C23
C14H17BO2S
MFCD11110552
2-benzo b thiophen-3-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-1-benzothiophen-3-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, benzo b thiophene-3-boronic acid pinacol ester, 2-benzo b thiophen-3-yl-4,4,5,5-tetramethyl-1,3,2 dioxaborolane, benzo b thiophene, 3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 2-benzo b thiophen-3-yl-4,4,5,5-tetramethyl-1,3,2-dioxaboro lane, 4,4,5,5-tetramethyl-2-benzo b thiophen-3-yl-1,3,2 dioxaborolane
2-(1-benzothiophen-3-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

Specifications

171364-86-6
MFCD11110552
2-benzo b thiophen-3-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-1-benzothiophen-3-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, benzo b thiophene-3-boronic acid pinacol ester, 2-benzo b thiophen-3-yl-4,4,5,5-tetramethyl-1,3,2 dioxaborolane, benzo b thiophene, 3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 2-benzo b thiophen-3-yl-4,4,5,5-tetramethyl-1,3,2-dioxaboro lane, 4,4,5,5-tetramethyl-2-benzo b thiophen-3-yl-1,3,2 dioxaborolane
B1(OC(C(O1)(C)C)(C)C)C2=CSC3=CC=CC=C23
260.158
260.16
Benzo[b]thiophene-3-boronic acid pinacol ester
C14H17BO2S
250 mg
LTSGSDOTQABJMA-UHFFFAOYSA-N
2-(1-benzothiophen-3-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
10658979
95%
Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only