missing translation for 'onlineSavingsMsg'
Learn More

Benzo[b]thiophene-3-boronic acid pinacol ester, 95%

CAS: 171364-86-6 | C14H17BO2S | 260.158 g/mol

Supplier:  Thermo Scientific Chemicals H5187603

 View more versions of this product

Catalog No. AAH5187603

Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Benzo[b]thiophene-3-boronic acid pinacol ester
C14H17BO2S
1 g
LTSGSDOTQABJMA-UHFFFAOYSA-N
2-(1-benzothiophen-3-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
10658979
95%
171364-86-6
MFCD11110552
2-benzo b thiophen-3-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-1-benzothiophen-3-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, benzo b thiophene-3-boronic acid pinacol ester, 2-benzo b thiophen-3-yl-4,4,5,5-tetramethyl-1,3,2 dioxaborolane, benzo b thiophene, 3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 2-benzo b thiophen-3-yl-4,4,5,5-tetramethyl-1,3,2-dioxaboro lane, 4,4,5,5-tetramethyl-2-benzo b thiophen-3-yl-1,3,2 dioxaborolane
B1(OC(C(O1)(C)C)(C)C)C2=CSC3=CC=CC=C23
260.158
260.16

Chemical Identifiers

171364-86-6
260.158
LTSGSDOTQABJMA-UHFFFAOYSA-N
10658979
B1(OC(C(O1)(C)C)(C)C)C2=CSC3=CC=CC=C23
C14H17BO2S
MFCD11110552
2-benzo b thiophen-3-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-1-benzothiophen-3-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, benzo b thiophene-3-boronic acid pinacol ester, 2-benzo b thiophen-3-yl-4,4,5,5-tetramethyl-1,3,2 dioxaborolane, benzo b thiophene, 3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 2-benzo b thiophen-3-yl-4,4,5,5-tetramethyl-1,3,2-dioxaboro lane, 4,4,5,5-tetramethyl-2-benzo b thiophen-3-yl-1,3,2 dioxaborolane
2-(1-benzothiophen-3-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only