missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzo[b]thiophene-3-boronic Acid (contains varying amounts of Anhydride), TCI America™
Specifications
Specifications
| Melting Point | 230°C |
| Quantity | 1 g |
| Synonym | benzothiophene-3-boronic acid, benzo b thiophen-3-ylboronic acid, benzo b thien-3-ylboronic acid, benzo b thiophene-3-boronic acid, thianaphthene-3-boronic acid, benzothiophen-3-ylboronic acid, 1-benzothiophen-3-yl-boranediol, 3-boronobenzo b thiophene, 1-benzothien-3-ylboronic acid, 3-borono-1-benzothiophene |
| Formula Weight | 178.01 |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Benzo[b]thiophene-3-boronic Acid (contains varying amounts of Anhydride) |
Safety and Handling
| TSCA | No |