accessibility menu, dialog, popup

UserName

HOBOHS

CAS RN 113893-08-6

IUPAC Name: (1-benzothiophen-3-yl)boronic acid
Synonyms: 1-benzothiophen-3-ylboronic acid
Molecular Weight (g/mol): 178.01
Molecular Formula: C8H7BO2S
InChi Key: QVANIYYVZZLQJP-UHFFFAOYSA-N
SMILES: OB(O)C1=CSC2=CC=CC=C12
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results