missing translation for 'onlineSavingsMsg'
Learn More

Benzo[b]thiophene-3-acetic acid, 98+%

Catalog No. AAL05855ME
Change view
Click to view available options
Quantity:
500 mg
2 g
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL05855ME 500 mg
AAL0585504 2 g
AAL0585509 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL05855ME Supplier Thermo Scientific Chemicals Supplier No. L05855ME
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1131-09-5 | C10H8O2S | 192.232 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1131-09-5
Molecular Formula C10H8O2S
Molecular Weight (g/mol) 192.232
MDL Number MFCD00051637
InChI Key VFZQJKXVHYZXMM-UHFFFAOYSA-N
Synonym benzo b thiophene-3-acetic acid, benzo b thiophen-3-yl acetic acid, 3-benzo b thienylacetic acid, acetic acid, 3-benzo b thienyl, 2-benzo b thiophen-3-yl acetic acid, 2-benzo b thiophen-3-ylacetic acid, benzothiophene-3-acetic acid, 2-1-benzothiophen-3-yl acetic acid, 3-benzo b thiopheneacetic acid
PubChem CID 70799
IUPAC Name 2-(1-benzothiophen-3-yl)acetic acid
SMILES C1=CC=C2C(=C1)C(=CS2)CC(=O)O

Specifications

Melting Point 108°C to 112°C
Quantity 500 mg
Beilstein 143357
Formula Weight 192.24
Percent Purity ≥98%
Chemical Name or Material Benzo[b]thiophene-3-acetic acid
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 214-461-2
RTECSNumber AF4800000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only