missing translation for 'onlineSavingsMsg'
Learn More

Benzo[b]thiophene-2-carboxamide, 97%

Catalog No. p-7082046
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1229203 1 g
AAA1229206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1229203 Supplier Thermo Scientific Chemicals Supplier No. A1229203
Only null left

CAS: 6314-42-7 | C9H7NOS | 177.221 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6314-42-7
Molecular Formula C9H7NOS
Molecular Weight (g/mol) 177.221
MDL Number MFCD00052505
InChI Key GYSCBCSGKXNZRH-UHFFFAOYSA-N
Synonym benzo b thiophene-2-carboxamide, thianaphthene-2-carboxamide, benzothiophene carboxamide, 2-benzo b thiophenecarboxamide, #
PubChem CID 237073
IUPAC Name 1-benzothiophene-2-carboxamide
SMILES C1=CC=C2C(=C1)C=C(S2)C(=O)N

Specifications

Melting Point 176°C to 178°C
Quantity 1 g
Beilstein 4467
Formula Weight 177.23
Percent Purity 97%
Chemical Name or Material Benzo[b]thiophene-2-carboxamide
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only